Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: IMKWVSDVDE VGDATSLPGC GEGASPGKAR RGPDGKANAS RKLCLPQPWM KTSGTHQKAS RRSLLGIRLM RSRNGRWEES KGSSWRRSMA WTSMKQVPAP TSTLKSHSRV
May act in concert with RAB3A in regulating aspects of synaptic vesicle membrane flow within the nerve terminal.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RAB15, member RAS onocogene family; Ras-related protein Rab-15; Ras-related protein Rab-15 isoform AN1; Ras-related protein Rab-15 isoform AN2; Ras-related protein Rab-15 isoform AN3
Gene Aliases: RAB15
UniProt ID: (Human) P59190
Entrez Gene ID: (Human) 376267
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support