Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MALRELRVCL LGDTGVGKSS IVWRFVEDSF DPNINPTIGA SFMTKTVQYQ NELHKFLIWD TAGQERFRAL APMYYRGSAA AIIVYDITKE ETFSTLKNWV KELRRHGPPN IVVAIAGNKC DLIDVREVME RDAKDYADSI HAIFVETSAK NAININELFI EISRRIPSTD ANLPSGGKGF KLRRQPSEPK RSCC
The protein encoded by this gene is a member of the RAB family of small GTPases. The GTP-bound form of the encoded protein has been shown to interact with early-endosomal antigen 1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP-binding protein RAB22A; MGC16770; Rab-22; Ras-related protein Rab-22A
Gene Aliases: RAB22; RAB22A
UniProt ID: (Human) Q9UL26
Entrez Gene ID: (Human) 57403
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support