Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSGQRVDVK VVMLGKEYVG KTSLVERYVH DRFLVGPYQN TIGAAFVAKV MSVGDRTVTL GIWDTAGSER YEAMSRIYYR GAKAAIVCYD LTDSSSFERA KFWVKELRSL EEGCQIYLCG TKSDLLEEDR RRRRVDFHDV QDYADNIKAQ LFETSSKTGQ SVDELFQKVA EDYVSVAAFQ VMTEDKGVDL GQKPNPYFYS CCHH (1-203 aa encoded by BC021263)
RAB24 may be involved in autophagy-related processes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Rab-16; Ras-related protein Rab-24
Gene Aliases: 6530406O07Rik; RAB24
UniProt ID: (Human) Q969Q5, (Mouse) P35290
Entrez Gene ID: (Human) 53917, (Mouse) 19336
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support