Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 150 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in mouse and rat.
Specificity of this antibody: RAB24.
RAB24 may be involved in autophagy-related processes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Rab-16; Ras-related protein Rab-24
Gene Aliases: 6530406O07Rik; RAB24
UniProt ID: (Human) Q969Q5, (Mouse) P35290
Entrez Gene ID: (Human) 53917, (Rat) 361208, (Mouse) 19336
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support