Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGNGTEEDY NFVFKVVLIG ESGVGKTNLL SRFTRNEFSH DSRTTIGVEF STRTVMLGTA AVKAQIWDTA GLERYRAITS AYYRGAVGAL LVFDLTKHQT YAVVERWLKE LYDHAEATIV VMLVGNKSDL SQAREVPTEE ARMFAENNGL LFLETSALDS TNVELAFETV LKEIFAKVSK QRQNSIRTNA ITLGSAQAGQ EPGPGEKRAC CISL (1-213 aa encoded by BC033322)
RAB proteins, such as RAB25, are members of the RAS superfamily of small GTPases that are involved in membrane trafficking. Members of the RAB11 subfamily, including RAB25, control the return of internalized membrane-associated moieties to the cell surface.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CATX-8; Ras-related protein Rab-25
Gene Aliases: CATX-8; CATX8; RAB11C; RAB25
UniProt ID: (Human) P57735
Entrez Gene ID: (Human) 57111
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support