Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%
RAB27B may be involved in targeting uroplakins to urothelial apical membranes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C25KG; Ras-related protein Rab-27B
Gene Aliases: 2310021G14Rik; B130064M09Rik; C25KG; RAB27B
UniProt ID: (Human) O00194, (Mouse) Q99P58
Entrez Gene ID: (Human) 5874, (Mouse) 80718
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support