Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: RDVKREEGEA FAREHGLIFM ETSAKTACNV EEAFINTAKE IYRKIQQGLF DVHNEANGIK IGPQQSISTS VGPSASQRNS RDIGSNSGCC
Members of the Rab protein family are nontransforming monomeric GTP-binding proteins of the Ras superfamily that contain 4 highly conserved regions involved in GTP binding and hydrolysis. Rab proteins are prenylated, membrane-bound proteins involved in vesicular fusion and trafficking.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP-binding protein RAB2B; RAS family, member RAB2B; Ras-related protein Rab-2B
Gene Aliases: 1500012D09Rik; 4930528G15Rik; A230002G14; D530043M21Rik; RAB2B
UniProt ID: (Human) Q8WUD1, (Mouse) P59279
Entrez Gene ID: (Human) 84932, (Mouse) 76338
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support