Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MMAIRELKVC LLGDTGVGKS SIVCRFVQDH FDHNISPTIG ASFMTKTVPC GNELHKFLIW DTAGQERFHS LAPMYYRGSA AAVIVYDITK QDSFYTLKKW VKELKEHGPE NIVMAIAGNK CDLSDIREVP LKDAKEYAES IGAIVVETSA KNAINIEELF QGISRQIPPL DPHENGNNGT IKVEKPTMQA SRRCC
Small GTP-binding proteins of the RAB family, such as RAB31, play essential roles in vesicle and granule targeting.[supplied by OMIM, Jul 2009].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP-binding protein Rab0; monomeric GTP-binding protein; Ras-related protein Rab-22B; Ras-related protein Rab-31
Gene Aliases: 1700093E07Rik; AI415285; Rab0; RAB22B; RAB31
UniProt ID: (Human) Q13636, (Rat) Q6GQP4, (Mouse) Q921E2
Entrez Gene ID: (Human) 11031, (Rat) 246324, (Mouse) 106572
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support