Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAGGGAGDP GLGAAAAPAP ETREHLFKVL VIGELGVGKT SIIKRYVHQL FSQHYRATIG VDFALKVLNW DSRTLVRLQL WDIAGQERFG NMTRVYYKEA VGAFVVFDIS RSSTFEAVLK WKSDLDSKVH LPNGSPIPAV LLANKCDQNK DSSQSPSQVD QFCKEHGFAG WFETSAKDNI NIEEAARFLV EKILVNHQSF PNEENDVDKI KLDQETLRAE NKSQCC (1-225 aa encoded by B C015061)
RAB32 acts as an A-kinase anchoring protein by binding to the type II regulatory subunit of protein kinase A and anchoring it to the mitochondrion. Also involved in synchronization of mitochondrial fission. Plays a role in the maturation of phagosomes that engulf pathogens, such as S.aureus and M.tuberculosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ras-related protein Rab-32
Gene Aliases: RAB32
UniProt ID: (Human) Q13637
Entrez Gene ID: (Human) 10981
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support