Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAQPILGHGS LQPASAAGLA SLELDSSLDQ YVQIRIFKII VIGDSNVGKT CLTFRFCGGT FPDKTEATIG VDFREKTVEI EGEKIKVQVW DTAGQERFRK SMVEHYYRNV HAVVFVYDVT KMTSFTNLKM WIQECNGHAV PPLVPKVLVG NKCDLREQIQ VPSNLALKFA DAHNMLLFET SAKDPKESQN VESIFMCLAC RLKAQKSLLY RDAERQQGKV QKLEFPQEAN SKTSCPC
The Ras-related superfamily of guanine nucleotide binding proteins includes the R-Ras, Rap, Ral/Rec and Rho/Rab subfamilies. Increasing data suggests an important role for Rab proteins in either endocytosis or in biosynthetic protein transport. The process of transporting newly synthesized proteins from the endoplasmic reticulum to various stacks of the Golgi complex and to secretory vesicles involves the movement of carrier vesicles and requires Rab protein function. Rab proteins are also an integral part of endocytic pathways. Rab 33B and Rab 33A share conserved effector domains, but share only 55.3% overall amino acid identity. This suggests that Rab 33B and Rab 33A may interact with similar effector and regulatory proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RAB 33A; Ras-related protein Rab-33A; Small GTP-binding protein S10
Gene Aliases: RAB33A; RABS10
UniProt ID: (Human) Q14088
Entrez Gene ID: (Human) 9363
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support