Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QAIIIVFNL NDVASLEHTK QWLADALKEN DPSSVLLFLT PAQYALMEKD ALQVAQEMKA EYWAVSSLTG ENVREFFFRV AALTFEANVL AELEKSGARR IGDVVRINSD DSNLYLTASK KKPTCCP (126-251 aa encoded by BC016841 )
Protein transport. Involved in the redistribution of lysosomes to the peri-Golgi region. Plays a role in the maturation of phagosomes that engulf pathogens, such as S. aureus and M. tuberculosis. Plays a role in the fusion of phagosomes with lysosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nine amino-acid residue-repeats; Ras-related protein Rab-34; Ras-related protein Rab-34, isoform NARR; Ras-related protein Rab-39; Ras-related protein Rah
Gene Aliases: NARR; RAB34; RAB39; RAH
UniProt ID: (Human) P0DI83
Entrez Gene ID: (Human) 83871
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support