Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: LVGTKKDLLS GAACEQAEAD AVHLAREMQA EYWSVSAKTG ENVKAFFSRV AALAFEQSVL QDLERQSSAR LQVGNGDLIQ MEGSPPETQE SKRPSSLGCC
Protein transport. Probably involved in vesicular traffic. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ras-related protein Rab-36; small GTP-binding protein Rab36
Gene Aliases: RAB36
UniProt ID: (Human) O95755
Entrez Gene ID: (Human) 9609
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support