Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Antibody detects endogenous levels of total RAB38.
May be involved in melanosomal transport and docking. Involved in the proper sorting of TYRP1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chocolate; Melanoma antigen NY-MEL-1; Rab-related GTP-binding protein; RAB38, member of RAS oncogene family; Ras-related protein Rab-38; red eyed dilution; ruby or red eyed dilution; small GTP binding protein
Gene Aliases: 2310011F14Rik; AU043391; cht; NY-MEL-1; R; RAB38; rrGTPbp; Ruby
UniProt ID: (Human) P57729, (Mouse) Q8QZZ8
Entrez Gene ID: (Human) 23682, (Rat) 252916, (Mouse) 72433
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support