Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MASATDSRYG QKESSDQNFD YMFKILIIGN SSVGKTSFLF RYADDSFTPA FVSTVGIDFK VKTIYRNDKR IKLQIWDTAG QERYRTITTA YYRGAMGFIL MYDITNEESF NAVQDWSTQI KTYSWDNAQV LLVGNKCDME DERVVSSERG RQLADHLGFE FFEASAKDNI NVKQTFERLV DVICEKMSES LDTADPAVTG AKQGPQLSDQ QVPPHQDCAC
Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has 4 known isoforms: 3A, 3B, 3C, and 3D. These proteins are expressed in highest concentrations in brain and endocrine tissues and involved in regulated exocytosis of hormones and neurotransmitters. Specifically, Rab 3s are thought to play a key role in presynaptic vesicle trafficking and priming.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C18A3.6; RAS-associated protein RAB3A; Ras-related protein Rab-3A; Ras-related small GTP binding protein 3A
Gene Aliases: RAB3; RAB3A
UniProt ID: (Pig) Q06AU3, (Human) P20336, (Rat) P63012, (Mouse) P63011
Entrez Gene ID: (Pig) 100144498, (Chicken) 107055371, (Human) 5864, (Rat) 25531, (Mouse) 19339
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support