Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Protein transport. Probably involved in vesicular traffic. Target protein may play a role in secretory granule function and exocytosis in pancreatic acinar cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain antigen RAB3B; Ras-related protein Rab-3B
Gene Aliases: 2610528C18Rik; RAB3B
UniProt ID: (Human) P20337, (Mouse) Q9CZT8
Entrez Gene ID: (Human) 5865, (Mouse) 69908
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support