Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MRHEAPMQMA SAQDARYGQK DSSDQNFDYM FKLLIIGNSS VGKTSFLFRY ADDSFTSAFV STVGIDFKVK TVFKNEKRIK LQIWDTAGQE RYRTITTAYY RGAMGFILMY DITNEESFNA VQDWSTQIKT YSWDNAQVIQ VGNKCDMEDE RVISTERGQH LGEQLGFEFF ETSAKDNINV KQTFERLVDI ICDKMSESLE TDPAITAAKQ NTRLKETPPP PQPNCAC
Rab proteins are low-molecular-weight GTP-binding proteins that form the largest branch of the Ras superfamily of GTPases. Located on the cytoplasmic face of organelles and vesicles, rab proteins are involved in intracellular membrane fusion reactions. Three membrane proteins, synaptosomal associated protein of 25 kDa (SNAP-25), synaptobrevin, and syntaxin, form the core of a ubiquitous membrane fusion machine that interacts with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and a-SNAP. Rab proteins, in coordination with the core fusion machinery and Munc-18, help to mediate vesicle docking and fusion. There exist over 40 Rab proteins that have been described in mammals. Rab 3C is found in brain, adrenal gland, and pituitary.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ras-related protein Rab-3C
Gene Aliases: RAB3C
UniProt ID: (Human) Q96E17
Entrez Gene ID: (Human) 115827
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support