Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Guanine nucleotide exchange factor (GEF) which may activate RAB8A and RAB8B. Promotes the exchange of GDP to GTP, converting inactive GDP-bound Rab proteins into their active GTP-bound form. Mediates the release of GDP from RAB8A and RAB8B but not from RAB3A or RAB5. Modulates actin organization and promotes polarized transport of RAB8A-specific vesicles to the cell surface. Together with RAB11A, RAB8A, the exocyst complex, PARD3, PRKCI, ANXA2, CDC42 and DNMBP promotes transcytosis of PODXL to the apical membrane initiation sites (AMIS), apical surface formation and lumenogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gene trap PAT 12; Rab-3A-interacting protein; RAB3A-interacting protein; RAB3IP; rab3ip {ECO:0000250|UniProtKB:Q96QF0}; rabin 3; Rabin-3; Rabin8; SSX2 interacting protein; SSX2-interacting protein
Gene Aliases: B230311A06; Gtpat12; RAB3IP; RABIN3; RABIN8
UniProt ID: (Human) Q96QF0, (Rat) Q62739, (Mouse) Q68EF0
Entrez Gene ID: (Human) 117177, (Rat) 29885, (Mouse) 216363
Molecular Function:
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support