Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSAPGSPDQA YDFLLKFLLV GDRDVGKSEI LESLQDGAAE SPYSHLGGID YKTTTILLDG QRVKLKLWDT SGQGRFCTIF RSYSRGAQGV ILVYDIANRW SFEGMDRWIK KIEEHAPGVP KILVGNRLHL AFKRQVPREQ AQAYAERLGV TFFEVSPLCN FNIIESFTEL ARIVLLRHRM NWLGRPSKVL SLQDLCCRTI VSCTPVHLVD KLPLPSTLRS HLKSFSMAKG LNARMMRGLS YSLTTSSTHK SSLCKVEIVC PPQSPPKNCT RNSCKIS
This gene encodes a member of the Rab40 subfamily of Rab small GTP-binding proteins that contains a C-terminal suppressors of cytokine signaling box.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein Rar-2; Ras-related protein Rab-40A; SOCS box containing protein RAR2A; SOCS box-containing protein RAR2A
Gene Aliases: RAB40A; RAR2; RAR2A
UniProt ID: (Human) Q8WXH6
Entrez Gene ID: (Human) 142684
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support