Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSALGSPVR AYDFLLKFLL VGDSDVGKGE ILASLQDGAA ESTYGHPAGI DYKTTTILLD GRRVKLQLWD TSGQGRFCTI FRSYSRGAQG VILVYDIANR WSFDGIDRWI KEIDEHAPGV PKILVGNRLH LAFKRQVPTE QAQAYAERLG VTFFEVSPLC NFNITESFTE LARIVLLRHG MDRLWRPSKV LSLQDLCCRA VVSCTPVHLV DKLPLPIALR SHLKSFSMAN GLNARMMHGG SYSLTTSSTH KRSSLRKVKL VRPPQSPPKN CTRNSCKIS (1-278 aa encoded by BC018039 )
Rab 40B is a 278 amino acid protein that belongs to the small GTPase superfamily and the Rab family. Rab 40B contains a SOCS box domain that mediates interactions with the Elongin BC complex, an adapter module in different E3 ubiquitin ligase complexes. Rab 40B shares similarity with a yeast protein, suggesting a role in regulation of secretory vesicles. The Rab 40B gene is conserved in chimpanzee, cow, mouse, rat, chicken and zebrafish, and maps to human chromosome 17q25.3. It has been suggested that there is a major gene locus on chromosome 17q25.3 for autosomal dominant moyamoya disease (MMD), which is an idiopathic steno-occlusive cerebrovascular disease that represents an important cause of stroke.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP-binding protein homologous to Saccharomyces cerevisiae SEC4; Protein Rar; Ras-related protein Rab-40B; sec4; SOCS box-containing protein RAR
Gene Aliases: RAB40B; RAR; SEC4L
UniProt ID: (Human) Q12829
Entrez Gene ID: (Human) 10966
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support