Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SVKNNCNVDL AFDTLADAIQ QALQQGDIKL EEGWGGVRLI HKTQIPRSPS RKQHSGPCQC
RAB42 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include membrane; GTP binding; GTPase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RAB42, member RAS homolog family; Ras-related protein Rab-42
Gene Aliases: RAB42
UniProt ID: (Human) Q8N4Z0
Entrez Gene ID: (Human) 115273
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support