Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GQERYHSMTR QLLRKADGVV LMYDITSQES FAHVRYWLDC LQDAGSDGVV ILLLGNKMDC EEERQVSVEA GQQLAQ
Antigen sequence identity: Mouse 83%; Rat 82%
RAB44 is a member of the RAS oncogene family of small GTPases that plays a role in intracellular vesicle trafficking and membrane fusion events. It is expressed in various tissues, including the brain, liver, and testis, and has been implicated in the regulation of autophagy and lysosomal function. However, its precise biological function and molecular mechanisms of action remain poorly understood.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RAS-like, family 13; Ras-related protein Rab-44; RASD family, member 3
Gene Aliases: dJ431A14.3; RAB44; RASD3; RASL13
UniProt ID: (Human) Q7Z6P3
Entrez Gene ID: (Human) 401258
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support