Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
RAB5B play a role in protein transport. RAB5B is probably involved in vesicular traffic.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ras-related protein Rab-5B
Gene Aliases: C030027M18Rik; RAB5B
UniProt ID: (Human) P61020, (Mouse) P61021
Entrez Gene ID: (Human) 5869, (Mouse) 19344
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support