Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The antigen shares 85% identity with RAB6C.
Specificity of this antibody: RAB6A and RAB6C.
GTPases of the RAB family bind to various effectors to regulate the targeting and fusion of transport carriers to acceptor compartments. This protein is located at the Golgi apparatus, which regulates trafficking in both a retrograde (from early endosomes and Golgi to the endoplasmic reticulum) and an anterograde (from the Golgi to the plasma membrane) directions. Myosin II is an effector of this protein in these processes. This protein is also involved in assembly of human cytomegalovirus (HCMV) by interacting with the cellular protein Bicaudal D1, which interacts with the HCMV virion tegument protein, pp150. Multiple alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq, Aug 2011]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Rab GTPase; Rab-6; RAB6, member RAS oncogene family; Rab6-like protein WTH3; RAB6B, member RAS oncogene family; Ras-related protein Rab-6A; Ras-related protein Rab-6B; Ras-related protein Rab-6C; small GTP-binding protein; small GTPase RAB6B
Gene Aliases: 2610028L11Rik; 9630015C03; AA419671; AI844641; C330006L04Rik; D9Bwg0185e; MNCb-1660; RAB6; RAB6A; RAB6B; RAB6C; RCO4-3; WTH3
UniProt ID: (Human) P20340, (Human) Q9NRW1, (Human) Q9H0N0, (Mouse) P35279, (Mouse) P61294
Entrez Gene ID: (Human) 5870, (Human) 51560, (Human) 84084, (Rat) 84379, (Mouse) 19346, (Mouse) 270192, (Rat) 363123
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support