Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MGSRDHLFKV LVVGDAAVGK TSLVQRYSQD SFSKHYKSTV GVDFALKVLQ WSDYEIVRLQ LWDIAGQERF TSMTRLYYRD ASACVIMFDV TNATTFSNSQ RWKQDLDSKL TLPNGEPVPC LLLANKCDLS PWAVSRDQID RFSKENGFTG WTETSVKENK NINEAMRVLI EKMMRNSTED IMSLSTQGDY INLQTKSSSW SCC
Rab GTPase key regulator in vesicle trafficking. Essential for maintaining the integrity of the endosome-trans-Golgi network structure. Together with LRRK2, plays a role in the retrograde trafficking pathway for recycling proteins, such as mannose 6 phosphate receptor (M6PR), between lysosomes and the Golgi apparatus in a retromer-dependent manner. Regulates neuronal process morphology in the intact central nervous system (CNS). May play a role in the formation of typhoid toxin transport intermediates during Salmonella enterica serovar Typhi (S.Typhi) epithelial cell infection. {ECO:0000269; PubMed:22042847, ECO:0000269; PubMed:24788816}.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Rab-7-like protein 1; RAB7, member RAS oncogene family-like 1; Ras-related protein Rab-29; Ras-related protein Rab-7L1
Gene Aliases: RAB29; RAB7L; RAB7L1
UniProt ID: (Human) O14966
Entrez Gene ID: (Human) 8934, (Mouse) 226422
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support