Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAGKSSLFKV ILLGDGGVGK SSLMNRYVTN KFDTQLFHTI GVEFLNKDLE VDGHFVTMQI WDTAGQERFR SLRTPFYRGS DCCLLTFSVD DSQSFQNLSN WKKEFIYYAD VKEPESFPFV ILGNKIDISE RQVSTEEAQA WCRDNGDYPY FETSAKDATN VAAAFEEAVR RVLATEDRSD HLIQTDTVNL HRKPKPSSSC C
Rab proteins are a family of Ras-like GTPases involved in intracellular compartment protein transport. Different members of the 40+ member rab family are responsible for docking and fusion of transport vesicles between different compartments within the cell. Rab 9 is required for trafficking mannose 6-phosphate receptor between the late endosome to trans-Golgi network (TGN). By facilitating receptor transport, rab 9 enables cells to efficiently recycle important cellular trafficking components. It is functionally necessary for rab 9, like other rab family members, to be prenylated by two 20-carbon geranylgeranyl groups at the C-terminus. Most prenylated rab 9 is membrane bound, however, 10-20% of the cellular pool of rab 9 is bound to GDP dissociation inhibitor-alpha (GDI-alpha) in the cytosol. GDI recycles prenylated, GDP bound rab 9 from their fusion targets back to their membranes of origin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RAB9, member RAS oncogene family; RAB9-like protein; Ras-related protein Rab-9A
Gene Aliases: RAB9; RAB9A
UniProt ID: (Human) P51151
Entrez Gene ID: (Human) 9367
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support