Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSGKSLLLK VILLGDGGVG KSSLMNRYVT NKFDSQAFHT IGVEFLNRDL EVDGRFVTLQ IWDTAGQERF KSLRTPFYRG ADCCLLTFSV DDRQSFENLG NWQKEFIYYA DVKDPEHFPF VVLGNKVDKE DRQVTTEEAQ TWCMENGDYP YLETSAKDDT NVTVAFEEAV RQVLAVEEQL EHCMLGHTID LNSGSKAGSS CC (1-201 aa encoded by BC093756)
Rab9b may be involved in the transport of proteins between the endosomes and the trans Golgi network.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Rab-9-like protein; Rab-9L; Ras-related protein Rab-9B; TRAB9
Gene Aliases: 9330195C02Rik; Rab-9L; RAB9B; RAB9L; RGD1562951
UniProt ID: (Human) Q9NP90, (Mouse) Q8BHH2
Entrez Gene ID: (Human) 51209, (Rat) 367915, (Mouse) 319642
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support