Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QKEFIYYADV KDPEHFPFVV LGNKVDKEDR QVTTEEAQTW CMENGDYPYL ETSAKDDTNV TVAFEEAVRQ VLAVEEQLEH CMLGHTIDLN SGSKAGSSCC
Rab9b may be involved in the transport of proteins between the endosomes and the trans Golgi network.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Rab-9-like protein; Rab-9L; Ras-related protein Rab-9B; TRAB9
Gene Aliases: Rab-9L; RAB9B; RAB9L
UniProt ID: (Human) Q9NP90
Entrez Gene ID: (Human) 51209
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support