Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: QQEDPIERFE RENRRLQEAN MRLEQENDDL AHELVTSKIA LRKDLDNAEE KADALNKELL MTKQKLIDAE EEKRRLEEES AQLKEMCRRE LDKAESEIKK NSSIIGDYKQ ICSQLSERLE KQQTANKVEI EKIRQKVDDC ERCREFFNKE GRVKGISSTK EVLDEDTDEE KETLKNQLRE MELELAQTKL QLVEAECKIQ DLEHHLGLAL NEVQAAKKTW FNRTLSSIKT ATGVQGKETC
RABGAP1 may act as a GTPase-activating protein of RAB6A and play a role in microtubule nucleation by centrosome. It may participate in a RAB6A-mediated pathway involved in the metaphase-anaphase transition.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GAP and centrosome-associated protein; Rab GTPase-activating protein 1; rab6 GTPase activating protein (GAP and centrosome-associated); Rab6 GTPase activating protein, GAPCenA; Rab6 GTPase-activating protein GAPCenA; rabgap1; RP11-123N4.2; TBC1 domain family, member 11
Gene Aliases: GAPCENA; HSPC094; Kiaa4104; mKIAA4104; RABGAP1; TBC1D11
UniProt ID: (Human) Q9Y3P9, (Mouse) A2AWA9
Entrez Gene ID: (Human) 23637, (Mouse) 227800
Molecular Function:
G-protein modulator
protein-binding activity modulator
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support