Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDDQPGEKEL VKRSQLDGEG DGPLSNQLSA SSTINPVPLV GLQKPEMSLP VKPGQGDSEA SSPFTPVADE DSVVFSKLTY LGCASVNAPR SEVEALRMMS ILRSQCQISL
RABGAP1 may act as a GTPase-activating protein of RAB6A and play a role in microtubule nucleation by centrosome. It may participate in a RAB6A-mediated pathway involved in the metaphase-anaphase transition.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GAP and centrosome-associated protein; Rab GTPase-activating protein 1; rab6 GTPase activating protein (GAP and centrosome-associated); Rab6 GTPase activating protein, GAPCenA; Rab6 GTPase-activating protein GAPCenA; RP11-123N4.2; TBC1 domain family, member 11
Gene Aliases: GAPCENA; HSPC094; RABGAP1; TBC1D11
UniProt ID: (Human) Q9Y3P9
Entrez Gene ID: (Human) 23637
Molecular Function:
G-protein modulator
protein-binding activity modulator
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support