Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FHCWDYRRF VATQAAVPPA EELAFTDSLI TRNFSNYSSW HYRSCLLPQL HPQPDSGPQG RLPEDVLLKE LELVQNAFFT DPNDQSAWFY HRWLLGRADP QDALRCLHVS RDEACLTVSF SRPLLVGSRM EILLLMVDDS PLIVEWRTPD GRNRPSHVWL CDLPAASLND QLPQHTFRVI WTAGDVQKEC VLLKGRQEGW CRDSTTDEQL FRCELSVEKS TVLQSELESC KELQELEPEN KWCLLTIILL MRALDPLLYE KETLQY (143-407 aa encoded by BC003093)
Catalyzes the transfer of a geranylgeranyl moiety from geranylgeranyl diphosphate to both cysteines of Rab proteins with the C-terminal sequence -XXCC, -XCXC and -CCXX, such as RAB1A, RAB3A, RAB5A and RAB7A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Geranylgeranyl transferase type II subunit alpha; Geranylgeranyl transferase type-2 subunit alpha; gunmetal; protein prenyltransferase alpha subunit repeat containing 3; Rab geranyl-geranyltransferase subunit alpha; Rab geranylgeranyl transferase, a subunit; Rab geranylgeranyltransferase alpha; Rab geranylgeranyltransferase subunit alpha; Rab geranylgeranyltransferase, alpha subunit; Rab GG transferase alpha; Rab GGTase alpha
Gene Aliases: Ggta; gm; PTAR3; RABGGTA
UniProt ID: (Human) Q92696, (Mouse) Q9JHK4, (Rat) Q08602
Entrez Gene ID: (Human) 5875, (Mouse) 56187, (Rat) 58983
Molecular Function:
acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support