Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MHGRLKVKTS EEQAEAKRLE REQKLKLYQS ATQAVFQKRQ AGELDESVLE LTSQILGANP DFATLWNCRR EVLQQLETQK SPEELAALVK AELGFLESCL RVNPKSYGTW HHRCWLLGRL PEPNWTRELE LCARFLEVDE RNFHCWDYRR FVATQAAVPP AEELAFTDSL ITRNFSNYSS WHYRSCLLPQ LHPQPDSGPQ GRLPEDVLLK ELELVQNAFF TDPNDQSAWF YHRWLLGRAD PQDALRCLHV SRDEACLTVS FSRPLLVGSR MEILLLMVDD SPLIVEWRTP DGRNRPSHVW LCDLPAASLN DQLPQHTFRV IWTAGDVQKE CVLLKGRQEG WCRDSTTDEQ LFRCELSVEK STVLQSELES CKELQELEPE NKWCLLTIIL LMRALDPLLY EKETLQYFQT LKAVDPMRAT YLDDLRSKFL LENSVLKMEY AEVRVLHLAH KDLTVLCHLE QLLLVTHLDL SHNRLRTLPP ALAALRCLEV LQASDNAIES LDGVTNLPRL QELLLCNNRL QQPAVLQPLA SCPRLVLLNL QGNPLCQAVG ILEQLAELLP SVSSVLT
Catalyzes the transfer of a geranylgeranyl moiety from geranylgeranyl diphosphate to both cysteines of Rab proteins with the C-terminal sequence -XXCC, -XCXC and -CCXX, such as RAB1A, RAB3A, RAB5A and RAB7A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Geranylgeranyl transferase type II subunit alpha; Geranylgeranyl transferase type-2 subunit alpha; protein prenyltransferase alpha subunit repeat containing 3; Rab geranyl-geranyltransferase subunit alpha; Rab geranylgeranyltransferase subunit alpha; Rab geranylgeranyltransferase, alpha subunit; Rab GG transferase alpha; Rab GGTase alpha
Gene Aliases: PTAR3; RABGGTA
UniProt ID: (Human) Q92696
Entrez Gene ID: (Human) 5875
Molecular Function:
acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support