Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine
RABGGTB catalyzes the transfer of a geranyl-geranyl moiety from geranyl-geranyl pyrophosphate to both cysteines in Rab proteins with an -XXCC, -XCXC and -CCXX C-terminal, such as RAB1A, RAB3A and RAB5A respectively.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Geranylgeranyl transferase type II subunit beta; Geranylgeranyl transferase type-2 subunit beta; GGTase-II-beta; Rab geranyl-geranyltransferase subunit beta; Rab geranylgeranyltransferase subunit beta; RAB geranylgeranyltransferase, beta subunit; Rab GG transferase beta; rab GGTase beta; Type II protein geranyl-geranyltransferase subunit beta
Gene Aliases: GGTB; RABGGTB
UniProt ID: (Human) P53611, (Mouse) P53612
Entrez Gene ID: (Human) 5876, (Mouse) 19352
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support