Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MGTPQKDVII KSDAPDTLLL EKHADYIASY GSKKDDYEYC MSEYLRMSGI YWGLTVMDLM GQLHRMNREE ILAFIKSCQH ECGGISASIG HDPHLLYTLS AVQILTLYDS INVIDVNKVV EYVKGLQKED GSFAGDIWGE IDTRFSFCAV ATLALLGKLD AINVEKAIEF VLSCMNFDGG FGCRPGSESH AGQIYCCTGF LAITSQLHQV NSDLLGWWLC ERQLPSGGLN GRPEKLPDVC YSWWVLASLK IIGRLHWIDR EKLRNFILAC QDEETGGFAD RPGDMVDPFH TLFGIAGLSL LGEEQIKPVN PVFCMPEEVL QRVNVQPELV S
RABGGTB catalyzes the transfer of a geranyl-geranyl moiety from geranyl-geranyl pyrophosphate to both cysteines in Rab proteins with an -XXCC, -XCXC and -CCXX C-terminal, such as RAB1A, RAB3A and RAB5A respectively.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Geranylgeranyl transferase type II subunit beta; Geranylgeranyl transferase type-2 subunit beta; GGTase-II-beta; Rab geranyl-geranyltransferase subunit beta; Rab geranylgeranyltransferase subunit beta; Rab geranylgeranyltransferase, beta subunit; Rab GG transferase beta; rab GGTase beta; Type II protein geranyl-geranyltransferase subunit beta
Gene Aliases: GGTB; RABGGTB
UniProt ID: (Human) P53611
Entrez Gene ID: (Human) 5876
Molecular Function:
acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support