Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MAEDKTKPSE LDQGKYDADD NVKIICLGDS AVGKSKLMER FLMDGFQPQQ LSTYALTLYK HTATVDGRTI LVDFWDTAGQ ERFQSMHASY YHKAHACIMV FDVQRKVTYR NLSTWYTELR EFRPEIPCIV VANKIDADIN VTQKSFNFAK KFSLPLYFVS AADGTNVVKL FNDAIRLAVS YKQNSQDFMD EIFQELENFS LEQEEEDVPD QEQSSSIETP SEEAASPHS
The RABL2B protein is a member of the RAB gene family which belongs to the RAS GTPase superfamily. RABL2B is located within a subtelomeric region of 22q13.3. Multiple alternatively spliced transcript variants encoding several different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HOX1B; Rab-like protein 2B
Gene Aliases: RABL2B
UniProt ID: (Human) Q9UNT1
Entrez Gene ID: (Human) 11158
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support