Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PCIPTLIGT PVKIGEGMLA DFVSQTSPMI PSIVVHCVNE IEQRGLTETG LYRISGCDRT VKELKEKFLR VKTVPLLSKV DDIHAICSLL KDFLRNLKEP LLTFRLNRAF MEAAEITDED NSIAAMYQAV GELPQANRDT LAFLMIHLQR VAQSPHTKMD VANLAKVFGP TIVAHAVPNP DPVTMLQDIK RQPKVVERLL SLPLEYWSQF MMVEQENIDP LHVIENSNAF STPQTPDIKV SLLGPVTTPE HQLLKTPSSS SLSQRVRSTL TKNTPRFGSK SKSATNLGRQ GNFFASPMLK (334-632 aa encoded by BC032754)
RAC, a small GTPase in the ras family, regulates the formation of membrane ruffles, lamellipodia and filopodia. It is involved in cytoskeletal actin organization and transformation. The p21 activated kinases (PAKs) are direct targets of active Rac and Cdc42 which can induce the assembly of polarized cytoskeletal structures when expressed in fibroblasts. RAC1 and RAC2 are also essential components of NADPH oxidase, the enzyme responsible for generating free radicals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AKT; CYK4; ID; Male germ cell RacGap; MGC99656; MgcRacGAP; PKB-ALPHA; PRKBA; protein CYK4 homolg; Protein CYK4 homolog; RAC; Rac GTPase-activating protein 1; RAC-ALPHA; racgap1
Gene Aliases: AI227039; AI327394; Band25; CYK4; gtl11; GTPase; HsCYK-4; ID-GAP; KIAA1478; MGCRACGAP; mKIAA1478; RACGAP1
UniProt ID: (Human) Q9H0H5, (Mouse) Q9WVM1
Entrez Gene ID: (Human) 29127, (Mouse) 26934, (Rat) 315298
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support