Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MQVTLKTLQ QQTFKIDIDP EETVKALKEK IESEKGKDAF PVAGQKLIYA GKILNDDTAL KEYKIDEKNF VVVMVTKPKA VSTPAPATTQ QSAPASTTAV TSSTTTTVAQ APTPVPALAP TSTPASITPA SATASSEPAP ASAAKQEKPA EKPAETPVAT SPTATDSTSG DSSRSNLFED ATSALVTGQS YENMVTEIMS MGYEREQVIA ALRASFNNPD RAVEYLLMGI PGDRESQAVV DPPQAASTGV PQSSAVAAAA ATTTATTTTT SSGGHPLEFL RNQPQFQQMR QIIQQNPSLL PALLQQIGRE NPQLLQQISQ HQEHFIQMLN EPVQEAGGQG GGGGGGSGGI AEAGSGHMNY IQVTPQEKEA IERLKALGFP EGLVIQAYFA CEKNENLAAN FLLQQNFDED (1-409 aa encoded by BC020973)
The protein encoded by this gene is one of two human homologs of Saccharomyces cerevisiae Rad23, a protein involved in the nucleotide excision repair (NER). This protein was found to be a component of the protein complex that specifically complements the NER defect of xeroderma pigmentosum group C (XP-c) cell extracts in vitro. This protein was also shown to interact with, and elevate the nucleotide excision activity of 3-methyladenine-DNA glycosylase (MPG), which suggested a role in DNA damage recognition in base excision repair. This protein contains an N-terminal ubiquitin-like domain, which was reported to interact with 26S proteasome, and thus this protein may be involved in the ubiquitin mediated proteolytic pathway in cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HR23B; p58; RAD23, yeast homolog of, B; RAD23b homolog; UV excision repair protein RAD23 homolog B; XP-C repair complementing complex 58 kDa; XP-C repair complementing protein; XP-C repair-complementing complex 58 kDa protein
Gene Aliases: 0610007D13Rik; AV001138; HHR23B; HR23B; Mhr23b; P58; RAD23B
UniProt ID: (Human) P54727, (Mouse) P54728, (Rat) Q4KMA2
Entrez Gene ID: (Human) 5887, (Mouse) 19359, (Rat) 298012
Molecular Function: damaged DNA-binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support