Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Five human RAD 51 homologues have been identified: XRCC2, XRCC3, RAD51B, RAD51C, and RAD51D. Each of these homologues interacts with one or more of the others, with all of the proteins involved in one complex or multiple smaller complexes. Rad51B plays a role in meiotic recombination and/or recombinational repair.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA repair protein RAD51 homolog 2; hREC2; MGC34245; R51H2; R51H2REC2hREC2; RAD51 homolog B; RAD51-like protein 1; Rad51B; RecA-like protein; recombination repair protein
Gene Aliases: R51H2; RAD51B; RAD51L1; REC2
UniProt ID: (Human) O15315
Entrez Gene ID: (Human) 5890
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support