Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In Western blot applications, this antibody detects a band at ~22kDa. A suggested positive control is Hela cell lysate.
PA5-19864 can be used with blocking peptide PEP-0004.
The protein encoded by this gene is a death domain (CARD/DD)-containing protein and has been shown to induce cell apoptosis. Through its CARD domain, this protein interacts with, and thus recruits, caspase 2/ICH1 to the cell death signal transduction complex that includes tumor necrosis factor receptor 1 (TNFR1A), RIPK1/RIP kinase, and numbers of other CARD domain-containing proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Caspase and RIP adapter with death domain; caspase and RIP adaptor with death domain; death adaptor molecule RAIDD; death domain containing protein CRADD; Death domain-containing protein CRADD; MGC9163; RIP-associated ICH1/CED3-homologous protein with death domain; RIP-associated protein with a death domain
Gene Aliases: CRADD; MRT34; RAIDD
UniProt ID: (Human) P78560
Entrez Gene ID: (Human) 8738
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support