Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: SLDVDNGNMY KSILVTSQDK APAVIRKAMD KHNLEEEEPE DYELLQILSD DRKLKIPENA NVFYAMNSTA NYDFVLKKRT FTKGVKVKHG ASSTLPRMKQ KGLKIAKGIF
Stimulates the dissociation of GDP from the Ras-related RalA and RalB GTPases which allows GTP binding and activation of the GTPases. Interacts and acts as an effector molecule for R-Ras, H-Ras, K-Ras, and Rap.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ral guanine nucleotide dissociation stimulator; Ral guanine nucleotide exchange factor; RalGDS; RalGEF
Gene Aliases: KIAA1308; RALGDS; RalGEF; RGDS; RGF
UniProt ID: (Human) Q9HCT1
Entrez Gene ID: (Human) 5900
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support