Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MREYKLVVLG SGGVGKSALT VQFVQGIFVE KYDPTIEDSY RKQVEVDCQQ CMLEILDTAG TEQFTAMRDL YMKNGQGFAL VYSITAQSTF NDLQDLREQI LRVKDTEDVP MILVGNKCDL EDERVVGKEQ GQNLARQWCN CAFLESSAKS KINVNEIFYD LVRQINRKTP VEKKKPKKKS CLLL
The product of this gene belongs to the family of RAS-related proteins. These proteins share approximately 50% amino acid identity with the classical RAS proteins and have numerous structural features in common. The most striking difference between RAP proteins and RAS proteins resides in their 61st amino acid: glutamine in RAS is replaced by threonine in RAP proteins. The product of this gene counteracts the mitogenic function of RAS because it can interact with RAS GAPs and RAF in a competitive manner. Two transcript variants encoding the same protein have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C21KG; G-22K; GTP-binding protein smg p21A; Ras-related protein Krev-1; Ras-related protein Rap-1A
Gene Aliases: C21KG; G-22K; KREV-1; KREV1; RAP1; RAP1A; SMGP21
UniProt ID: (Human) P62834
Entrez Gene ID: (Human) 5906
Molecular Function: small GTPase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support