Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MIEKMQGSRM DEQRCSFPPP LKTEEDYIPY PSVHEVLGRE GPFPLILLPQ FGGYWIEGTN HEITSIPETE PLQSPTTKVK LECNPTARIY RKHFLGKEHF NYYSLDAALG HLVFSLKYDV IGDQEHLRLL LRTKCRTYHD VIPISCLTEF PNVVQMAKLV CEDVNVDRFY PVLYPKASRL IVTFDEHVIS NNFKFGVIYQ KLGQTSEEEL FSTNEESPAF VEFLEFLGQK VKLQDFKGFR GGLDVTHGQT GTESVYCNFR NKEIMFHVST KLPYTEGDAQ QLQRKRHIGN DIVAVVFQDE NTPLVPDMIA SNFLHAYVVV QAEGGGPDGP LYKVSVTARD DVPFFGPPLP DPAVFRKGPE FQEFLLTKLI NAEYACYKAE KFAKLEERTR AALLETLYEE LHIHSQSMMG LGGDEDKMEN GSGGGGFFES FKRVIRSRSQ SMDAMGLSNK KPNTVSTSHS GSFAPNNPDL AKAAGISLIV PGKSPTRKKS GPFGSRRSSA IGIENIQEVQ EKRESPPAGQ KTPDSGHVSQ EPKSENSSTQ SSPEMPTTKN RAETAAQRAE ALKDFSRSSS SASSFASVVE ETEGVDGEDT GLESVSSSGT PHKRDSFIYS TWLEDSVSTT SGGSSPGPSR SPHPDAGKLG DPACPEIKIQ LEASEQHMPQ LGC
Rap1GAP specifically stimulates GTP hydrolytic activity of the monomeric G protein Rap1. Physical interaction between G alpha z, a member of the Gi family of trimeric G proteins, and Rap1GAP blocks the ability of regulators of G protein signaling to stimulate GTP hydrolysis of the alpha subunit, and also attenuates the ability of activated G alpha z to inhibit adenylyl cyclase. Rap1GAP is expressed in the brain, kidney and pancreas and may act as a signal integrator to coordinate and/or integrate Gz signaling and Rap1 signaling in cells. A novel isoform of Rapl GTPase-activating protein, designated Rap1GAPII, binds specifically to G alpha z. Stimulation of the Gi-coupled M2 Muscarinic receptor translocates Rap1GAPII from the cytosol to the membrane and decreases the amount of GTP-bound Rap1, resulting in the activation of ERK/MAPK.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Rap1 GTPase-activating protein 1; Rap1GAP
Gene Aliases: KIAA0474; RAP1GA1; RAP1GAP; RAP1GAP1; RAP1GAPII; RAPGAP
UniProt ID: (Human) P47736
Entrez Gene ID: (Human) 5909
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support