Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MFDCMDVLSV SPGQILDFYT ASPSSCMLQE KALKACFSGL TQTEWQHRHT AQSIETQSTS SEELVPSPPS PLPPPRVYKP CFVCQDKSSG YHYGVSACEG
Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HBV-activated protein; hepatitis B virus activated protein; Nuclear receptor subfamily 1 group B member 2; RAR beta; RAR-b; RAR-beta; RAR-Beta2; RAR-Beta4; RAR-epsilon; retinoic acid; retinoic acid receptor; Retinoic acid receptor beta; retinoic acid receptor, beta polypeptide
Gene Aliases: HAP; MCOPS12; NR1B2; RARB; RARbeta1; RRB2
UniProt ID: (Human) P10826
Entrez Gene ID: (Human) 5915
Molecular Function:
C4 zinc finger nuclear receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support