Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDVLVSECS ARLLQQEEEI KSLTAEIDRL KNCGCLGASP NLEQLQEENL KLKYRLNILR KSLQAERNKP TKNMINIISR LQEVFGHAIK AAYPDLENPP LLVTPSQQAK FGDYQCNSAM GISQMLKTKE QKVNPREIAE NITKHLPDNE CIEKVEIAGP GFINVHLRKD FVSEQLTSLL VNGVQLPALG E (1-190 aa encoded by BC000528)
The fidelity of protein synthesis requires efficient discrimination of amino acid substrates by aminoacyl-tRNA synthetases. ArgRS (Arginyl-tRNA synthetase), also known as RARS or DALRD1, belongs to the class-I aminoacyl-tRNA synthetase family that includes the related proteins, LeuRS, ValRS and IleRS. These proteins are large monomeric proteins and play a major role in catalyzing the aminoacylation of tRNA by their cognate amino acid. ArgRS localizes to the cytoplasm and exists as a monomer but can also associate with other tRNA synthetases and auxiliary proteins to form a multisubunit complex. In the presence of ATP, arginine (Arg) and tRNA, ArgRS joins Arg to tRNA(Arg) at its synthetic active site. Two cytoplasmic forms of ArgRS have been described in mammals, differing by the addition of a 73 amino acid sequence that is required for ArgRS assembly into the multisubunit complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arginine tRNA ligase 1, cytoplasmic; Arginine--tRNA ligase, cytoplasmic; Arginyl-tRNA synthetase; arginyl-tRNA synthetase, cytoplasmic; ArgRS; MGC8641
Gene Aliases: 2610011N19Rik; 2610037E21Rik; AL033339; ArgRS; AW985894; DALRD1; HLD9; RARS; RARS1
UniProt ID: (Human) P54136, (Rat) P40329, (Mouse) Q9D0I9
Entrez Gene ID: (Human) 5917, (Rat) 287191, (Mouse) 104458
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support