Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LKAREAEAWE EPTELLGLPS ALAGMQDLRL EGALHLLLAQ QQLRARARAG SVRPRGQPTP GEMLPSLPVG SDSEGPENEG TRAALAAAGL SGGRH
Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 55%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RBBP8 N-terminal-like protein
Gene Aliases: C20orf151; RBBP8NL
UniProt ID: (Human) Q8NC74
Entrez Gene ID: (Human) 140893
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support