Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MASPSKAVI VPGNGGGDVT THGWYGWVKK ELEKIPGFQC LAKNMPDPIT ARESIWLPFM ETELHCDEKT IIIGHSSGAI AAMRYAETHR VYAIVLVSAY TSDLGDENER ASGYFTRPWQ WEKIKANCPY IVQFGSTDDP FLPWKEQQEV ADRLETKLHK FTDCGHFQNT EFHELITVVK SLLKVPA (1-186 aa encoded by BC015938)
RBBP9 may play a role in the transformation process due to its capacity to confer resistance to the growth-inhibitory effects of TGF-beta1 through interaction with retinoblastoma and the subsequent displacement of E2F-1.The protein encoded by this gene is a retinoblastoma binding protein that may play a role in the regulation of cell proliferation and differentiation. Two alternatively spliced transcript variants of this gene with identical predicted protein products have been reported, one of which is a nonsense-mediated decay candidate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B5T overexpressed gene protein; B5T-overexpressed gene protein; MGC9236; Protein BOG; Putative hydrolase RBBP9; RBBP-10; RBBP-9; retinoblastoma binding protein 9; Retinoblastoma-binding protein 10; Retinoblastoma-binding protein 9; retinoma-binding protein 9; Serine hydrolase RBBP9
Gene Aliases: BOG; RBBP10; RBBP9
UniProt ID: (Human) O75884, (Rat) O88350, (Mouse) O88851
Entrez Gene ID: (Human) 10741, (Pig) 100738866, (Rat) 29459, (Mouse) 26450
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support