Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: PIRDRRGSLE KSQGDKRDRK NSASAERDRK HRTTAPTEGK SPLKKEDRSD GSAPSTSTAS SKLKSPSQKQ DGGTAPVASA SPKLCLAWQG MLLLKNSNFP SNMHLLQGDL
May function as an mRNA export factor, stimulating export and expression of RTE-containing mRNAs which are present in many retrotransposons that require to be exported prior to splicing. High affinity binding of pre-mRNA to RBM15 may allow targeting of the mRNP to the export helicase DBP5 in a manner that is independent of splicing-mediated NXF1 deposition, resulting in export prior to splicing. May be implicated in HOX gene regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: one twenty two protein; one twenty-two; One-twenty two protein 1; RNA-binding motif protein 15; RNA-binding protein 15
Gene Aliases: OTT; OTT1; RBM15; SPEN
UniProt ID: (Human) Q96T37
Entrez Gene ID: (Human) 64783
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support