Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPMLDIRPG LIPQAPGPRF PLIQPGIPPQ RGIPPPSVLD SALHPPPRGP FPPGDIFSQP ERPFLAPGRQ SVDNVTNPEK RIPLGNDNIQ QEGDRDYRFP PIETRESISR PPPVDVRDVV GRPIDPREGP GRPPLDGRDH FGRPPVDIRE NLVRPGIDHL GRRDHFGFNP EKPWGHRGDF DEREHRVLPV YGGPKGLHEE RGRFRSGNYR FDPRSGPWNR GFGQEVHRDF DDRRRPWERQ RDRDDRDFDF CREMNGNRLG RDRIQNTWVP PPHARVFDYF EGATSQRKGD NVPQVNGENT ERHAQPPPIP VQNDPELYEK LTSSNEINKE KSDTVADIES EPVVESTETE GT (921-1271 aa encoded by BC070071)
Regulates the p53 pathway to control the expansion growth of digestive organs. Target protein binds the repetitive C-terminal domain (CTD) of RNA polymerase II; may act with other CTD binding proteins to link transcription and pre-mRNA processing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC5L complex-associated protein 7; CTD-binding SR-like protein rA8; protein SCAF8; putative RNA-binding protein 16; RNA binding motif protein 16; RNA-binding motif protein 16; SR-like CTD-associated factor 8; SR-related and CTD-associated factor 8; SR-related CTD-associated factor 8
Gene Aliases: A630086M08Rik; A930036P18Rik; AA408306; AI447644; AI448652; AU015035; CCAP7; KIAA1116; mKIAA1116; RBM16; SCAF8
UniProt ID: (Human) Q9UPN6, (Mouse) Q6DID3
Entrez Gene ID: (Human) 22828, (Mouse) 106583
Molecular Function:
RNA processing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support