Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Phosphoinositide-binding protein which associates with both cell and endoplasmic reticulum (ER) membranes. Can bind to the ER membrane protein VAPA and recruit VAPA to plasma membrane sites, thus linking these intracellular compartments. The ORP3-VAPA complex stimulates RRAS signaling which in turn attenuates integrin beta-1 (ITGB1) activation at the cell surface. With VAPA, may regulate ER morphology. Has a role in regulation of the actin cytoskeleton, cell polarity and cell adhesion. Binds to phosphoinositides with preference for PI(3,4)P2 and PI(3,4,5)P3. Also binds 25-hydroxycholesterol and cholesterol.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2810480G15Rik; RNA-binding motif protein 28; RNA-binding protein 28
Gene Aliases: ANES; RBM28
UniProt ID: (Human) Q9NW13
Entrez Gene ID: (Human) 55131
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support