Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
RBM33 gene ontology annotations related to this gene include RNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp434D1319; DKFZp686F102; hypothetical protein MGC20460; MGC20460; proline rich 8; Proline-rich protein 8; RNA-binding motif protein 33; RNA-binding protein 33
Gene Aliases: 3200001K10Rik; 6430512A10Rik; AA386890; PRR8; RBM33
UniProt ID: (Human) Q96EV2, (Mouse) Q9CXK9
Entrez Gene ID: (Human) 155435, (Mouse) 381626
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support