Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKRVNSCVK SDEHVLEELE TEGERQLKSL LQHQLDTSVS IEECMSKKES FAPGTMYKPF GKEAAGTMTL SQFQTLHEKD QETASLRELG LNETEILIWK SHVSGEKKTK LRATPEAIQN RLQDIEERIS ERQRILCLPQ RFAKSKQLTR REMEIEKSLF QGADRHSFLK ALYYQDEPQK KNKGDPMNNL ESFYQEMIMK KRLEEFQLMR GEPFASHSLV SATSVGDSGT AESPSLLQDK GKQAAQGKGP SLHVANVIDF SPEQCWTGPK KLTQPIEFVP EDEIQRNRLS EEEIRKIPMF SSYNPGEPNK VLYLKNLSPR VTERDLVSLF ARFQEKKGPP IQFRMMTGRM RGQAFITFPN KEIAWQALHL VNGYKLYGKI LVIEFGKNKK QRSNLQATSL ISCATGSTTE ISGS (1-413 aa encoded by BC006986)
May bind RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RNA-binding motif protein 41; RNA-binding protein 41
Gene Aliases: BB131279; D330023I21Rik; RBM41
UniProt ID: (Human) Q96IZ5, (Mouse) Q8JZV4
Entrez Gene ID: (Human) 55285, (Mouse) 237073, (Rat) 680581
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support